ContactUsLinkComponent
Hamster Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal dete ction antibody, Streptavidin-HRP, recombinant hamster Interferon gamma ELISA standard, and standard reconstitution bu 1 * 576 Tests
Codice catalogo ANTIA270372-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Hamster Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal dete ction antibody, Streptavidin-HRP, recombinant hamster Interferon gamma ELISA standard, and standard reconstitution bu 1 * 576 Tests
Codice catalogo ANTIA270372-696
Nome del fornitore:  ANTIBODIES.COM