Human IL-12/IL-23 p40 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-AP, recombinant human IL-12 p70 ELISA standard, and standard reconstitution buffer. This ma 1 * 1.920 Tests
Codice catalogo ANTIA270401-2096
Nome del fornitore: ANTIBODIES.COM