Human IL-12 p70 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant human IL-12 p70 ELISA standard (lyophilized), and standard reconstitution buffer 1 * 576 Tests
Codice catalogo ANTIA270486-696
Nome del fornitore: ANTIBODIES.COM