ContactUsLinkComponent
Cotton Rat Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal d etection antibody, Streptavidin-HRP, recombinant cotton rat Interferon gamma ELISA standard, and standard reconstitut 1 * 1.920 Tests
Codice catalogo ANTIA270465-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Cotton Rat Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal d etection antibody, Streptavidin-HRP, recombinant cotton rat Interferon gamma ELISA standard, and standard reconstitut 1 * 1.920 Tests
Codice catalogo ANTIA270465-2096
Nome del fornitore:  ANTIBODIES.COM