Human TGF beta 1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant human LAP ELISA standard, and standard reconstitution buffer. This matched anti 1 * 1.920 Tests
Codice catalogo ANTIA270503-2096
Nome del fornitore: ANTIBODIES.COM