ContactUsLinkComponent
Human IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant human IL-1a ELISA standard, and standard reconstitution buffer. This matched an 1 * 576 Tests
Codice catalogo ANTIA270482-696
Nome del fornitore:  ANTIBODIES.COM
New Product
Human IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant human IL-1a ELISA standard, and standard reconstitution buffer. This matched an 1 * 576 Tests
Codice catalogo ANTIA270482-696
Nome del fornitore:  ANTIBODIES.COM