Human IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-22 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
ANTIA270491-696