Monkey TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human TNF alpha ELISA standard, and standard reconstitution buffer. This matched 1 * 1.920 Tests
Codice catalogo ANTIA270430-2096
Nome del fornitore: ANTIBODIES.COM