Bovine IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antib ody, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched anti 1 * 1.920 Tests
Codice catalogo ANTIA270374-2096
Nome del fornitore: ANTIBODIES.COM