ContactUsLinkComponent
Human MCP1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human CCL2 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 1.920 Tests
Codice catalogo ANTIA325333-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Human MCP1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, recombinant human CCL2 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 1.920 Tests
Codice catalogo ANTIA325333-2096
Nome del fornitore:  ANTIBODIES.COM