ContactUsLinkComponent
Rat Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detectio n antibody, Streptavidin-AP, recombinant rat Interferon gamma ELISA standard, and standard reconstitution buffer. Thi 1 * 1.920 Tests
Catalog # ANTIA325351-2096
Supplier:  ANTIBODIES.COM
New Product
Rat Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detectio n antibody, Streptavidin-AP, recombinant rat Interferon gamma ELISA standard, and standard reconstitution buffer. Thi 1 * 1.920 Tests
Catalog # ANTIA325351-2096
Supplier:  ANTIBODIES.COM