Human Epstein Barr Virus EBNA1 IgG ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro quantitative determina tion of human EBV Nuclear Antigen / EBNA1 IgG in serum and plasma. 1 * 96 Tests
Codice catalogo ANTIA324324-96
Nome del fornitore: ANTIBODIES.COM