Monkey IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Codice catalogo ANTIA325375-2096
Nome del fornitore: ANTIBODIES.COM