Mouse IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant mouse IL-1ß ELISA standard, and standard reconstitution buffer. This match 1 * 576 Tests
Codice catalogo ANTIA325339-696
Nome del fornitore: ANTIBODIES.COM