ContactUsLinkComponent
Porcine IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant porcine IL-5 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Codice catalogo ANTIA325355-2096
Nome del fornitore:  ANTIBODIES.COM
New Product
Porcine IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant porcine IL-5 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 1.920 Tests
Codice catalogo ANTIA325355-2096
Nome del fornitore:  ANTIBODIES.COM