Hamster IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, native Syrian hamster IgG ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 576 Tests
Codice catalogo ANTIA325362-696
Nome del fornitore: ANTIBODIES.COM