Bovine TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant bovine TNF alpha ELISA standard, and standard reconstitution buffer. This matche 1 * 1.920 Tests
Catalog # ANTIA325323-2096
Supplier: ANTIBODIES.COM