Bovine uPA Receptor/U-PAR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine uPA Receptor/U-PAR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Codice catalogo ANTIA4143-48
Nome del fornitore: ANTIBODIES.COM