ContactUsLinkComponent
Acrylamide-ES ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determina tion of Acrylamide-ES in food samples such as potato/corn chips, french fries, cereals, and breads. 1 * 96 Tests
Codice catalogo ANTIA327173-96
Nome del fornitore:  ANTIBODIES.COM
New Product
Acrylamide-ES ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determina tion of Acrylamide-ES in food samples such as potato/corn chips, french fries, cereals, and breads. 1 * 96 Tests
Codice catalogo ANTIA327173-96
Nome del fornitore:  ANTIBODIES.COM