ContactUsLinkComponent
Chicken CCL4/MIP-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of chicken CCL4/MIP-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA4368-48
Supplier:  ANTIBODIES.COM
New Product
Chicken CCL4/MIP-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of chicken CCL4/MIP-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA4368-48
Supplier:  ANTIBODIES.COM