Viresolve® Pro Device
Viresolve® pro device, Parvovirus removal filter, H×L×W: 13,56×18,62×9,22 cm, 0,22 m²
MILXVPMD103NB1
New Product
: Merck
The Viresolve® pro solution provides a comprehensive, flexible template solution for viral clearance within mAb and recombinant protein bioproduction. This proven viral clearance solution delivers the highest levels of retention assurance and productivity across a broad range of feed stream characteristics.
Fully disposable single-use fluid path
Devices 100% tested with air/water diffusion and binary gas test
Shield/shield H 100% aerosol tested
Viresolve pro modus devices are typically used for filtration of pilot to mid-scale batch volumes of proteins. The Viresolve® Pro Device Modus 1,2 and 1,3 offers a filtration area of 0,07 and 0.22 m² and has a primary use in pilot/small-volume processing.
- Superficie di filtrazione:0,22 m²
- Descrizione:Viresolve® pro device, Parvovirus removal filter, H×L×W: 13,56×18,62×9,22 cm



