ContactUsLinkComponent
Aniline [CAS:62-53-3] 100 ug/ml in Methanol CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator and protected from light 1 * 1 mL
Codice catalogo ACSDP804190
Nome del fornitore:  CPACHEM
New Product
Aniline [CAS:62-53-3] 100 ug/ml in Methanol CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator and protected from light 1 * 1 mL
Codice catalogo ACSDP804190
Nome del fornitore:  CPACHEM