Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Rack with 96 Sample Tubes for Use with SampleJet

Rack with 96 Sample Tubes for Use with SampleJet

103,5 mm, 1,7 mm, Yes, 0,20 mm
Codice catalogo BRUKZ106462
New Product
Nome del fornitore:   BRUKER DALTRONIK
Rack with 96 Sample Tubes for Use with SampleJet
Codice catalogo BRUKZ106462
Nome del fornitore:  BRUKER DALTRONIK
New Product
Rack with 96 Sample Tubes for Use with SampleJet

103,5 mm, 1,7 mm, Yes, 0,20 mm

Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Rack with 96 coded tubes with caps designed for high throughput use with SampleJet. 60 µm camber.

Bruker caps are crafted from high-quality Orgalloy® plastic, offering good resistance to a variety of chemicals and NMR solvents such as CDCl3, MeOD, and DMSO. However, it is advisable to prevent direct contact between the sample solution and the cap.

Specifiche
  • Tappo incluso:
    Yes
  • Ø est:
    1,7 mm
  • Spessore parete:
    0,20 mm
  • Lunghezza:
    103,5 mm