EPQPYEEIPIYL, A PHOSPHOPEPTIDE, IS A SRC HOMOLOGY 2 (SH2) DOMAIN LIGAND. EPQPYEEIPIYL ACTIVATES SRC FAMILY MEMBERS (E.G. LCK, HCK, F YN) BY BINDING TO SH2 DOMAIN
Codice catalogo MDEXHY-P3279-10MG
Nome del fornitore: MEDCHEMEXPRESS
Frequently Bought Together