2,4,6-Trimethoxybenzaldehyde 98%
Bottle
Codice catalogo APOSOR10592-25G
Nome del fornitore: Apollo Scientific
2,4,6-Trimethoxybenzaldehyde 98%
Codice catalogo APOSOR10592-25G
Nome del fornitore: Apollo Scientific
Numero CAS: 830-79-5
Vai a
Product Details & Documents
Usage: Organic building block/ reagent.
- Numero CAS:830-79-5
- Formula:C₁₀H₁₂O₄
- Numero MDL:MFCD00003313
- Molecular Weight:196,19988 g/mol
Specifiche
Specifiche
- Tipo di conf.:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



