Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
4-(2-Hydroxyethoxy)benzaldehyde 95%

4-(2-Hydroxyethoxy)benzaldehyde 95%

Bottle
Katalognr. APOSOR922883-100G
Reizend
New Product
Anbieter:   Apollo Scientific
4-(2-Hydroxyethoxy)benzaldehyde 95%
Katalognr. APOSOR922883-100G
Anbieter:  Apollo Scientific
New Product
CAS Nummer:  22042-73-5
4-(2-Hydroxyethoxy)benzaldehyde 95%

Bottle

CAS Nummer:  22042-73-5
Preis
SFr. 612.00
Einloggen für Verfügbarkeit
1 * 100 g
Product Details & Documents
  • CAS Nummer:22042-73-5
  • Formel:C₉H₁₀O₃
  • MDL-Nummer:MFCD00191450
  • Molekulargewicht:166.1739 g/mol
  • Schmelzender Platingehalt:40 °C
  • Lagertemperatur:Ambiente
Technische Daten

Technische Daten

  • Tipo di conf.:
    Bottle

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.