ContactUsLinkComponent
Potassium Hydroxide 0.1M (0.1N) CRM, ISO 17034 and ISO 17025 Conservation 12 months Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1 L
  ACSDZ14571065.1L
 :  CPACHEM
New Product
Potassium Hydroxide 0.1M (0.1N) CRM, ISO 17034 and ISO 17025 Conservation 12 months Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1 L
  ACSDZ14571065.1L
 :  CPACHEM