Potassium Hydroxide 0.5M (0.5N) CRM, ISO 17034 and ISO 17025 Conservation 24 months Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1 L
Codice catalogo ACSDZ14571066.1L
Nome del fornitore: CPACHEM