ETHYL 2-BROMO-2,2-DIFLUOROACETATE 1 * 100 g
Catalog # AMBDA608634-100G
Supplier: AMBEED
Frequently Bought Together
Metanolo ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade per LC-MS, Supelco®
Supplier: Merck
1-Butanolo ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Glicerina 85% Ph. Eur.
Supplier: VWR Chemicals
Cicloesano ≥99.5%, PESTINORM® per analisi dei pesticidi
Supplier: VWR Chemicals
Diclorometano ≥99.5% stabilizzato, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acqua, LiChrosolv® per cromatografia, per LC-MS, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS per analisi, Supelco®
Supplier: Merck
Etanolo 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Metanolo ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetraidrofurano, extra dry over Molecular Sieve 99.5% stabilizzato, AcroSeal®
Supplier: Thermo Fisher Scientific