ContactUsLinkComponent
16 components: Cu 100mg/l Sn 100mg/l Fe 200mg/l Cr 50mg/l Ni 50mg/l Mn 50mg/l Zn 50mg/l Pb 20mg/l As 20mg/l Al 20mg/l Ti 20mg/l Cd 10mg/l Hg 10mg/l Co 10mg/l Sb 10mg/l V 10mg/l in HNO3 10% Conservation 12 months. Store u 1 * 100 mL
Catalog # CMPLRM116328L1
New Product
16 components: Cu 100mg/l Sn 100mg/l Fe 200mg/l Cr 50mg/l Ni 50mg/l Mn 50mg/l Zn 50mg/l Pb 20mg/l As 20mg/l Al 20mg/l Ti 20mg/l Cd 10mg/l Hg 10mg/l Co 10mg/l Sb 10mg/l V 10mg/l in HNO3 10% Conservation 12 months. Store u 1 * 100 mL
Catalog # CMPLRM116328L1