ContactUsLinkComponent
Wide Mouth Bottle 1000 mL, HDPE (conforms to FDA 177.1520 & USP Class VI). Cap PP screwed on (conforms to FDA 177.1520 & USP Class VI). For transporting samples, suitable for powders and liquids. Made in a Class 100,000 cleanroom. Height: 170 mm. Dia 1 * 24 pezzi
Codice catalogo SAMP8126H-1000
Nome del fornitore:  Sampling Systems
New Product
Wide Mouth Bottle 1000 mL, HDPE (conforms to FDA 177.1520 & USP Class VI). Cap PP screwed on (conforms to FDA 177.1520 & USP Class VI). For transporting samples, suitable for powders and liquids. Made in a Class 100,000 cleanroom. Height: 170 mm. Dia 1 * 24 pezzi
Codice catalogo SAMP8126H-1000
Nome del fornitore:  Sampling Systems