[EN]ISOFIELD HORIZON C/ALL W/HOOD M 1 * 20 pezzi
Catalog # ISOF80020M
Supplier: ISOFIELD IRELAND
New Product
[EN]ISOFIELD HORIZON C/ALL W/HOOD M 1 * 20 pezzi
Catalog # ISOF80020M
Supplier: ISOFIELD IRELAND
New Product
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Puntali per pipette, Finntip™
Supplier: Thermo Fisher Scientific
Acido trifluoroacetico, Uvasol® per spettroscopia, Supelco®
Supplier: Merck
Siringhe monouso, 3 pezzi, HENKE-JECT®
Supplier: HENKE SASS WOLF
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Etanolo 96% vol Ph. Eur.
Supplier: VWR Chemicals
Etanolo assoluto ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, HiPerSolv CHROMANORM®, grado gradiente per HPLC
Supplier: VWR Chemicals
Water standards, CRM, Karl Fischer titration, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
[EN]ISOFIELD HORIZON C/ALL W/HOOD XXL 1 * 20 pezzi
Supplier: ISOFIELD IRELAND