[1,3-BIS(DIPHENYLPHOSPHINO)PROPANE]PALLA 1 * 5 g
Catalog # AMBDA555253-5G
Supplier: AMBEED
Frequently Bought Together
Alcol iso-amilico, TECHNICAL
Supplier: VWR Chemicals
(R)-(+)-Pulegone per sintesi, Sigma-Aldrich®
Supplier: Merck
N-Metil-2-pirrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimetil pentano ≥99% (by GC), CHROMASOLV™ per analisi dei pesticidi, Riedel-de Haën™
Supplier: Honeywell Chemicals
Polivinil alcool, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Metanolo ≥99.9%, CHROMASOLV™ per analisi GC capillare, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimetilformammide 99+%, extra puro
Supplier: Thermo Fisher Scientific
[EN]N-HEPTANE 95% CHROMAR HPLC 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
Acetone ≥99.5% (by GC), liquido USP, BP, Ph. Eur., puro
Supplier: PanReac AppliChem
Metanolo ≥99.8%, HiPerSolv CHROMANORM®, grado isocratico per HPLC
Supplier: VWR Chemicals
Acetone ≥99.5% ACS
Supplier: VWR Chemicals
1-Pentanolo 99+% ACS
Supplier: Thermo Fisher Scientific
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals