Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Sartobind® Rapid A Labs

Sartobind® Rapid A Labs

Sartobind® Rapid A lab, 203×158×78 mm
Codice catalogo SARTSBLRA025EL-A
   Produzione a basso impatto
Nome del fornitore:   Sartorius
Sartobind® Rapid A Labs
Codice catalogo SARTSBLRA025EL-A
Nome del fornitore:  Sartorius
   Produzione a basso impatto
Sartobind® Rapid A Labs
Sartobind® Rapid A Labs

Sartobind® Rapid A lab, 203×158×78 mm

Price
SFr. 727.00
Log in per disponibilità
1 * 4 ST
Product Details & Documents

To affinity and beyond: Fast, flexible and future-proof antibody purification.

  • The fastest matrix. High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes

  • Productivity boost. Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity
  • Plug and play. Collapse your workflow. Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration
  • Flexibility built in. Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system
  • Process-ready technology. Seamless scalability. Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production

Sartobind® Rapid A Lab incorporates the state-of-the-art in membrane affinity chromatography (AC) technology, offering fast, flexible and future-proof antibody purification. It is an ideal solution for antibody research, as well as downstream process development for novel biologics before starting clinical and commercial production.

Bed volume: 0,5 ml

Ligand: Protein A

Membrane support: Polyethylene terephthalate (PET)

Specifiche
  • Descrizione:
    Sartobind® Rapid A lab, 203×158×78 mm
  • Seq:
    2
  • Cat. No.:
    SARTSBLRA025EL-A
  • Environmentally Preferable:
    Produzione a basso impatto
  • Certificazione ambientale:
    Renewable Energy Certificate
Accessories

Informazioni sul prodotto

  • Codice catalogo
    Disponibilità
    Price
  • Sartobind® Lab LC system adapter kit, 60×80×10 mm
    Sartobind® Lab LC system adapter kit, 60×80×10 mm
    SARTSBLAAU01-1
    Log in per disponibilità
    1 * 1 ST - SFr. 31.00

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..