Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Sartobind® Rapid A Labs

Sartobind® Rapid A Labs

Sartobind® Rapid A lab, 125×120×62 mm
Codice catalogo SARTSBLRA025EL-1
   Produzione a basso impatto
Nome del fornitore:   Sartorius
Sartobind® Rapid A Labs
Codice catalogo SARTSBLRA025EL-1
Nome del fornitore:  Sartorius
   Produzione a basso impatto
Sartobind® Rapid A Labs
Sartobind® Rapid A Labs

Sartobind® Rapid A lab, 125×120×62 mm

Price
SFr. 211.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

To affinity and beyond: Fast, flexible and future-proof antibody purification.

  • The fastest matrix. High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes

  • Productivity boost. Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity
  • Plug and play. Collapse your workflow. Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration
  • Flexibility built in. Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system
  • Process-ready technology. Seamless scalability. Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production

Sartobind® Rapid A Lab incorporates the state-of-the-art in membrane affinity chromatography (AC) technology, offering fast, flexible and future-proof antibody purification. It is an ideal solution for antibody research, as well as downstream process development for novel biologics before starting clinical and commercial production.

Bed volume: 0,5 ml

Ligand: Protein A

Membrane support: Polyethylene terephthalate (PET)

Specifiche
  • Descrizione:
    Sartobind® Rapid A lab, 125×120×62 mm
  • Seq:
    1
  • Cat. No.:
    SARTSBLRA025EL-1
  • Environmentally Preferable:
    Produzione a basso impatto
  • Certificazione ambientale:
    Renewable Energy Certificate
Accessories

Informazioni sul prodotto

  • Codice catalogo
    Disponibilità
    Price
  • Sartobind® Lab LC system adapter kit, 60×80×10 mm
    Sartobind® Lab LC system adapter kit, 60×80×10 mm
    SARTSBLAAU01-1
    Log in per disponibilità
    1 * 1 ST - SFr. 31.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.