SC2930-H GENE FRAGMENTS: NAME:6F_HPEP4NOSTOP_9R SEQUENCE LENGTH:285 BP ADAPTER: YES, DELIVERY FORM: FREEZE DRIED FORMAT: SINGLE TUB ES YIELD: 500NG 1 * 1 ST
Catalog # MSPPOFU5931743G0-5
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together