ContactUsLinkComponent
SomnoSuite Starter Kit for mice, includes (2) 5mL glass syringes, low profile anesthesia mask, (1) .5L sliding top induction chamber , 2-pk charcoal canisters 1 * 1 ST
Codice catalogo MSPPSOMNO-MSEKIT
Nome del fornitore:  MARKET SOURCE PART PROCESS
SomnoSuite Starter Kit for mice, includes (2) 5mL glass syringes, low profile anesthesia mask, (1) .5L sliding top induction chamber , 2-pk charcoal canisters 1 * 1 ST
Codice catalogo MSPPSOMNO-MSEKIT
Nome del fornitore:  MARKET SOURCE PART PROCESS
Frequently Bought Together