POF-L2.5-1SMA-FTC-SU OPTICAL FIBER FOR USE WITH FTC-SU+SST SENSORS POF POLYMER OPTICAL FIBER L2.5 LENGTH OF OPTICAL FIBER IS 2.5 MET ER 1 SMA CONNECTOR DISTAL END IS POLISHED FIBER FTC SU FOR USE WITH FTC-SU AND SST SENSORS 1 * 1 ST
MSPP200002254
Frequently Bought Together