b.safe Laboratory Flasks
500 ml, GLS 80
Codice catalogo BOHLR100-43
Nome del fornitore: Bohlender
Vai a
Product Details & Documents
Laboratory flask made of borosilicate glass.
Includes screw cap and spout made of PP
Specifiche
- Filettatura:GLS 80
- Capacità:500 ml
- Seq:02