b.safe Disc Sensor Holders
Disc sensor holder, 81×130 mm, Waste cap, GL 45
BOHLR649-45
Made of PE, height-adjustable bracket for a b.safe disc sensor (catalogue number BOHLR640-05).
Simple attachment of the b.safe disc sensor by hanging the holder into the b.safe waste cap
- Position of the sensor can be stepless adjusted in height for level monitoring
- Use in containers and canisters made of non-conductive materials
The disc sensor is clipped into the corresponding holder, the cable guide prevents kinks on the sensor cable. After having put the holder on the PTFE insert of the waste cap, the disc sensor can be positioned freely at the required height. The adjusted height corresponds to the fill level to start the alarm.
- Per:Waste cap, GL 45
- Descrizione:Disc sensor holder, 81×130 mm
- Seq:01