Evolution™ UV/VIS Spectrophotometers
Empower the analysis and bring versatility and productivity to your lab with the evolution uv-vis spectrophotometers. With next-generation INSIGHT™ PRO software, the instrument is always up to date and ready for the next sample challenge.
- Double-beam geometry
- Xenon flash lamp requires no warm-up time allowing instant measurements
- Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices to complete the analysis
For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting edge instrumentation and a wide selection of accessories, the evolution systems work with you to perform experiments the way you want - for the results you need.
INSIGHT PRO Software - Sophistication Simplified
Personalize your home screen for easy navigation and convenience. Workbook tools save time and ensure accurate results. CUE scripting allows creation of customized user interface and simplified workflow. Create a dedicated workflow to guide for complex assays prevents costly sampling and measurement errors. New user interface reduces training time: prompting software defines clear step-by-step workflow and is easy to teach and learn. Always improving – weather it is user inputs or application requirements, or new regulations, Insight Pro is constantly improving with quick upgrades to deliver greater capabilities and efficiencies for the lab needs.
INSIGHT PRO with Security Software
Ensures integrity of scientific data required by pharmaceutical, biotech companies and academics working on FDA regulated studies and helps your lab stay compliant. Comply with US FDA 21 CFR Part 11 – A) Control user account access, apply digital signatures, allows to implement controls, electronic audit trails for a reliable electronic documentation. B) Database storage ensures to maintain electronic records for FDA submissions.
Comply with current 2019 USP, EP and JP pharmacopeia requirements: Validate your system: Includes methods to run IQ/OQ verification test to evaluate instrument performance to run within manufacturer’s specifications and satisfy current pharmacopeia requirements. Option to network single or multiple instruments – Insight pro security software on a central computer to control privileges and view audit. Logs across multiple evolution instruments in different labs.
Optimized Engineering Delivers Ultimate Performance
Double-beam with one nm spectral bandwidth for precision measurements. Application focused beam geometry technology, on the evolution one and evolution one plus models, enables the best performance from microcells, solid samples and fiber optic probes. Xenon flash lamp requires no warm-up time allowing instant measurements. Precision monochromator drive delivers fast scanning data collection and wavelength accuracy.
Assurance of System Performance
The thermo scientific uv validator package provides support for system qualification and validation activities. Documentation facilitates compliance of your system with the requirements of FDA, GxP, ISO 9001:2000 and ISPE 2001 guidelines and regulations. Mercury lamp accessory is available to perform instrument calibration. Thermo scientific insight pro security software provides complete tools to meet 21CFR part 11 requirements.
Accessories information: Accessories for all the sampling needs. Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete the analysis. Choice of Peltier single cell holder or a smart 8-cell Peltier system for temperature control. Easy software integration gives complete control of the measurements.
Documents
- Modello:Evolution One Plus USP/EP standards
- Descrizione:Evolution one USP/EP standards pharma bundle includes, evolution one UV-Vis spectrometer, INSIGHT Pro with security software, validator documentation A4 paper version, USP and EP UV standards set, USP and EP visible standards set
- Seq:04
- Sorgente luminosa:—
- Peso:—
- Dimensioni:—
- Sistema ottico:Double beam
- Range spettrale:—
- Intervalli dati (scansione):—
- Intervallo fotometrico:—
- Cat. No.:SPET840-370600
- Elettrico:—
- Rivelatore:—
- Velocità di scansione:—
- Display:—
- Banda passante spettrale:1 nm
Accessories
- DescrizioneModelloPer
- SPET222-269400Evolution ISA-220 integrating sphere, Single beam optical design, 1-2 nm spectral bandwidth, 220 – 1100 nm wavelength range, 10×10 mm silicon photodiode detector, 10, 5, 2, 1,0, 0,5, 0,2, 0,1 nm data interval, 9×5×7, 1 lbs.Transmission/reflectance measurementAccessory for evolution 220 spectrometers1 * 1 ST - SFr. 14’420.00
- DescrizioneModelloPer
- SPET699-1312008-cell peltier system, Operates on all voltages and 50/60 Hz, temperature range 5...100 °C with 0.1 °C precision, 24×17×14, 35 lbs.Temp/time based kinetic applicationAccessory for evolution spectrometers1 * 1 ST - SFr. 14’950.00
- DescrizioneModelloPer
- SPET840-219000Smart thermostatted 7-cell changer, 15×9×10, 16 lbs., 0...95 °C temp. range—Accessory for evolution spectrometers1 * 1 ST - SFr. 5’500.00
- DescrizioneModelloPer
- SPET840-326000Qualification filter kit for UV-VIS, 381×254×177 mm, 1,36 kg, 240 - 640 nm wavelength rangeCertified standardKit to qualifify spectrophotometer for UV-Visible region1 * 1 ST - SFr. 2’730.00
- DescrizioneModelloPer
- DescrizioneModelloPer
- SPET222-269500Cuvette holder for ISA-220 integrating sphere—Accessory for evolution 220 spectrometers1 * 1 ST - SFr. 1’350.00



