Orders LinkContactUsLinkComponent
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
TableWorx performance work table, type 304 stainless steel work surface, legs, and leg mounts, stainless steel three-sided frame, model: TW-SU-304-S, 762,0×1524,0 mm
Codice catalogo METOTWS3060SU304S
Nome del fornitore:   Metro International
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Codice catalogo METOTWS3060SU304S
Nome del fornitore:  Metro International
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
TableWorx performance work table, type 304 stainless steel work surface, legs, and leg mounts, stainless steel three-sided frame, model: TW-SU-304-S, 762,0×1524,0 mm
Price
SFr. 3’350.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
TableWorx is a truly modular platform of performance work tables designed to allow you to customise your perfect solution.
All stainless series TableWorx performance work tables feature a 16-gauge, Type 304 stainless steel work surface reinforced with 16-gauge hat channels. The sound deadened work surface features 90° turned down edges on all four sides. All sizes are available with an optional 51 mm stainless steel backsplash, which is formed into the work surface.
All stainless series TableWorx performance work tables feature type 304 stainless steel legs and leg mounts. A type 304 stainless steel three-sided frame allows for equipment to be nested underneath the table, and is also ideal for seated use of the work table.
Stationary work table legs include stainless steel leveling bullet feet with 29 mm range of adjustability. Mobile-ready versions are also available, and can accept any Metro 25 mm stem caster. Please note that casters are NOT included with mobile-ready TableWorx models and must be ordered separately.
Elevate your efficiency to new levels with the addition of TableWorx productivity riser. Risers can be easily added to any TableWorx work table. A full array of overshelves, wire grids, and accessories can be added to maximise every inch of space, making you more comfortable and productive than ever before. Risers can be mounted at the rear or center of the work table.
Specifiche
  • Descrizione:
    TableWorx performance work table, type 304 stainless steel work surface, legs, and leg mounts, stainless steel three-sided frame, model: TW-SU-304-S, 762,0×1524,0 mm
  • Seq:
    06