Screw Cap GL 40
GL 40, nero, PP
Codice catalogo RIXIU8S1001401800
Nome del fornitore: BERLIN PACKAGING RIXIUS GMBH
Vai a
Product Details & Documents
Screw cap made of black PP with ALU laminated PE foam insert for thread GL 40.
- Height: 14,4 mm
- Weight: 6 g
The PP screw caps makes it very popular for packaging food and cosmetics, among other things.
Specifiche
- Filettatura:GL 40
- Colore:nero
- Materiale:PP
- Seq:02
- Cat. No.:RIXIU8S1001401800