Orders LinkContactUsLinkComponent
Container, EUROBOX
Container, EUROBOX
414 L, with two handles, 1200×800×510 mm
Codice catalogo ZARG40709
Nome del fornitore:   ZARGES LEICHTBAU
Container, EUROBOX
Codice catalogo ZARG40709
Nome del fornitore:  ZARGES LEICHTBAU
Container, EUROBOX
Container, EUROBOX
414 L, with two handles, 1200×800×510 mm
Price
1 190.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Aluminium, with lid.
  • Lid with stainless steel hinges and holding straps
  • Blue stacking corners ensure safe stacking
  • Corner beads for additional dimensional stability
Eurobox has high resistance against corrosion due to stainless steel fittings. The peripheral foamed seal in the lid protects against dust and splash water.
Specifiche
  • Tipo:
    with two handles
  • Capacità:
    414 L
  • Seq:
    04
  • L×P×A:
    1200×800×510 mm