0,00 SFr.
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
KINGFISHER BIOTECH CYNOMOLGUS MONKEY GM-CSF RECOMBINANT PROTEIN 25 MICROGRAM. 1 * 25 µG
KINFRP1185Y-025
:
Kingfisher Biotech
KINGFISHER BIOTECH CYNOMOLGUS MONKEY GM-CSF RECOMBINANT PROTEIN 25 MICROGRAM. 1 * 25 µG
KINFRP1185Y-025
:
Kingfisher Biotech
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Pipette sierologiche, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Contenitori per campioni dritti con tappo a vite
: VWR Collection
VWR®, Provette e strip per PCR, 0,2 ml
: VWR Collection
VWR®, Pipette Pasteur, Standard Line
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
Pipette sierologiche, Stripette™
: Corning
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF