Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Fristads® 4083 High Visibility Softshell Jackets, Class 2

Fristads® 4083 High Visibility Softshell Jackets, Class 2

blu scuro/giallo, L, Regular
Codice catalogo KWIN125024171L
Nome del fornitore:   FRISTADS KANSAS
Fristads® 4083 High Visibility Softshell Jackets, Class 2
Codice catalogo KWIN125024171L
Nome del fornitore:  FRISTADS KANSAS
Fristads® 4083 High Visibility Softshell Jackets, Class 2

blu scuro/giallo, L, Regular

Price
SFr. 197.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

High visibility softshell jacket made of a windproof and water-repellent stretch material suitable for most of the year.

  • Windproof, water repellent and breathable
  • Stretch material; Functional pockets
  • Fleece inside
  • Full-length front zip with inside placket
  • Chest pocket with zip
  • Two front pockets with zip; Inside pocket with zip
  • ID-card pocket

Adjustable cuffs with velcro fastening, adjustable hem with drawstring and extended back.

 : EN ISO 20471 class 2. OEKO-TEX® certified.

Specifiche
  • Taglia dell'abbigliamento:
    L
  • Vestibilità del capo:
    Regular
  • Colore:
    blu scuro/giallo
  • Seq:
    04
  • Tessuto:
    100% polyester
  • Cat. No.:
    KWIN125024171L
  • Sleeve or leg length:
    Long
  • Risk area:
    HiVis
  • Peso:
    305 g/m²
  • Modello:
    4083
  • Applicazioni per abiti:
    Industrial|Outdoor
  • Tipo di chiusura:
    Con chiusura a zip
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.