Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
GemCell™ U.S. Human Serum AB | Avantor

GemCell™ U.S. Human Serum AB

U.S. human serum AB, gamma-irradiated, Bottle
Katalognr. GEMB100-512G-100
Anbieter:   GEMINI BIO-PRODUCTS
GemCell™ U.S. Human Serum AB
Katalognr. GEMB100-512G-100
GemCell™ U.S. Human Serum AB
GemCell™ U.S. Human Serum AB

U.S. human serum AB, gamma-irradiated, Bottle

Preis
655.00 SFr.
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents

GemCell™ human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from source plasma AB. All donor units are tested for viral markers and found to be non-reactive.

  • 0,1 µm sterile-filtered
  • Gamma irradiation available
Technische Daten
  • Descrizione:
    U.S. human serum AB, gamma-irradiated
  • Seq:
    02
  • Tipo di conf.:
    Bottle
  • Cat. No.:
    GEMB100-512G-100
  • Origine:
    US
  • Temperatura di conservazione:
    Store frozen at −20...−10 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.