Orders LinkContactUsLinkComponent
Axygen® LTS Compatible Pipette Tips | Avantor
Axygen® LTS Compatible Pipette Tips
LTS compatible, 200 µl, +, -, 50,29 mm, Clear, 10 racks, 96 each
Katalognr. 613-7419
Anbieter:   Corning
Axygen® LTS Compatible Pipette Tips
Katalognr. 613-7419
Anbieter:  Corning
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips
LTS compatible, 200 µl, +, -, 50,29 mm, Clear, 10 racks, 96 each
Preis
SFr. 81.40
Einloggen für Verfügbarkeit
1 * 960 ST
Product Details & Documents
The new Axygen® LTS compatible pipette tips are ergonomically designed to reduce discomfort and are optimised to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimised to work with LTS pipettors enables less force to be used during pipetting.
Zertifizierung: ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
Technische Daten
  • Confezione:
    10 racks, 96 each
  • Sterili:
    +
  • Volume:
    200 µl
  • Lunghezza:
    50,29 mm
  • Seq:
    13
  • Stile punta:
    LTS compatible
  • Colore puntale:
    Clear
  • Filtro:
    -