Orders LinkContactUsLinkComponent
Accessori
Accessori
Milli-Q® SP 240 prefilter module, for pretreatment of potable tap water to feed the pure RO water production station, includes a pressure regulator, requires one power cord
Codice catalogo MILLZSR240PFT0
Nome del fornitore:   Merck
Accessori
Codice catalogo MILLZSR240PFT0
Nome del fornitore:  Merck
Accessori
Milli-Q® SP 240 prefilter module, for pretreatment of potable tap water to feed the pure RO water production station, includes a pressure regulator, requires one power cord
Price
SFr. 1’700.00
Log in per disponibilità
1 * 1 ST
Specifiche
  • Descrizione:
    Milli-Q® SP 240 prefilter module, for pretreatment of potable tap water to feed the pure RO water production station, includes a pressure regulator, requires one power cord
  • Seq:
    50