Alpha Sterile Kits
Product Details & Documents
Sterile coverall with mandarin collar, hood, and overboots.
- Manufactured in a facility operating under ISO 9001:2015 quality management system
- Processed in a NEBB certified ISO Class 5 cleanroom
Mandarin collar coverall:
GMP Grade A and B, ISO Class 4 and 5 cleanroom compatible
Two-layer mandarin collar provides secure fit at neck area
Zip flap with double-sided tape prevents ingress and egress of particles
Low-linting, anti-static fabric suitable for chemo handling
Elasticated thumb loops
Back elastic to prevent bellows effect
Hood:
Latex-free enclosed elastic face opening for comfortable fit
Extra-long yoke for added protection
Three panel hood design to minimise skin exposure
Overboots:
Extra-long knee height
Two-ties for added security
Anti-slip PU sole
Confezione: 1 set (hood/coverall/boots) per sealed inner bag. 1 inner bag per sealed outer bag. 40 outer bags per carton liner with twist tie (S to XXL). 35 sets per carton (3XL).
Specifiche
- Taglia dell'abbigliamento:M
- Confezione:40/pack
- Sterili:+
- Colore:bianco
- Seq:02
- Cat. No.:ISOF80551M
- Classe di cleanroom:10/ISO 4
- Antistatico:+
- Tipo di chiusura:Con chiusura a zip